Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AIM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584779
Description
Immunogen sequence: LGCGAASGTP SGILYEPPAE KEQKVLIQSV SCTGTEDTLA QCEQEEVYDC.
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
Specifications
AIM | |
Polyclonal | |
Unconjugated | |
Cd5l | |
1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; CD5L; CT2; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; igM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; Spalpha; Sp-alpha; UNQ203/PRO229 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43866 | |
Cd5l | |
Recombinant protein corresponding to Human CD5L. Recombinant protein control fragment (Product #RP-103668). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction