Learn More
Invitrogen™ AIM Recombinant Rabbit Monoclonal Antibody (38)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CD5L/Aim.
Specifications
Specifications
| Antigen | AIM |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 38 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Cd5l |
| Gene Accession No. | Q9QWK4 |
| Gene Alias | 1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; CD5L; CT2; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; igM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; Spalpha; Sp-alpha; UNQ203/PRO229 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.