Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIM1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5122257
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122257 100 μL
PIPA5122258 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122257 Supplier Invitrogen™ Supplier No. PA5122257
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AIM1 Polyclonal antibody specifically detects AIM1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CRYBG1
Gene Accession No. Q9Y4K1
Gene Alias absent in melanoma 1; absent in melanoma 1 protein; AIM1; beta/gamma crystallin domain-containing protein 1; beta-gamma crystallin domain containing 1; CRYBG1; ST4; suppression of tumorigenicity 4 (malignant melanoma)
Gene Symbols CRYBG1
Host Species Rabbit
Immunogen Recombinant Absent In Melanoma 1 (AIM1), Asp346-Met576 (Accession Q9Y4K1).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.