Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AIM1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556214
Description
Immunogen sequence: TNWLTYSGTQ RVGSLYPIKQ RRVYFRLWNA ALGGFLAVPD HVEDMKAGRV VVADPQAGGS CIWYYEDGLL KNQMAPTMSL QVIGPPSPGS KVVLW Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.
Specifications
| AIM1L | |
| Polyclonal | |
| Unconjugated | |
| CRYBG2 | |
| absent in melanoma 1-like; absent in melanoma 1-like protein; AIM1L; beta/gamma crystallin domain-containing protein 2; beta-gamma crystallin domain containing 2; CRYBG2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1P7 | |
| CRYBG2 | |
| Recombinant protein corresponding to Human AIM1L. Recombinant protein control fragment (Product #RP-94468). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction