Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AIM1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556215
Description
Immunogen sequence: PEISLFSEEG LKGEQVKLTE ALKNSQGLEK PLQVASATVS AGLWLLYPKP LFEDTPYILE PGEYPTSEAW GTSDPSVGSL KPMRL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.

Specifications
AIM1L | |
Polyclonal | |
Unconjugated | |
CRYBG2 | |
absent in melanoma 1-like; absent in melanoma 1-like protein; AIM1L; beta/gamma crystallin domain-containing protein 2; beta-gamma crystallin domain containing 2; CRYBG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55057 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N1P7 | |
CRYBG2 | |
Recombinant protein corresponding to Human AIM1L. Recombinant protein control fragment (Product #RP-94469). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction