Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM1L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988814
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8814 Supplier Bioss Supplier No. BS9093R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen AIM1L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CRYBG2
Gene Accession No. Q8N1P7
Gene Alias absent in melanoma 1-like; Absent in melanoma 1-like protein; AIM1L; Beta/gamma crystallin domain-containing protein 2; beta-gamma crystallin domain containing 2; CRYBG2
Gene Symbols CRYBG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AIM1L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55057
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.