Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIM2 Antibody (10M2B3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280566
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.Specifications
AIM2 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 0.3-2ug/ml | |
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
Mouse | |
Protein G purified | |
RUO | |
9447 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot | |
10M2B3 | |
Unconjugated | |
PBS | |
AIM2 | |
This antibody was raised against recombinant human AIM2 protein | |
0.1 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction