Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIM2 Antibody (10M2B3), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | AIM2 |
---|---|
Clone | 10M2B3 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.3-2ug/ml |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227355SS
![]() |
Novus Biologicals
NBP227355SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227355
![]() |
Novus Biologicals
NBP227355 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.Specifications
AIM2 | |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
AIM2 | |
IgG2a κ | |
Protein G purified |
10M2B3 | |
Western Blot 0.3-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
O14862 | |
9447 | |
This antibody was raised against recombinant human AIM2 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title