Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIM2 Antibody (10M5G5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227354MFV450
Description
AIM2 Monoclonal antibody specifically detects AIM2 in Human samples. It is validated for Western BlotSpecifications
AIM2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
9447 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot | |
10M5G5 | |
Western Blot | |
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
This antibody was raised against AIM2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction