Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIM2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299853AF532
Description
AIM2 Polyclonal specifically detects AIM2 in Human samples. It is validated for ELISA.Specifications
AIM2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
9447 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 532 | |
absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
Produced in rabbits immunized with purified, recombinant AIM2 (NP_004824.1; Met1-Thr343). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction