Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIM2 Monoclonal Antibody (10M2B3)
GREENER_CHOICE

Catalog No. PIMA516259
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516259 Supplier Invitrogen™ Supplier No. MA516259
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIM2 Monoclonal Antibody for Western Blot

AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIM2
Applications Western Blot
Classification Monoclonal
Clone 10M2B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene AIM2
Gene Accession No. O14862
Gene Alias Absent in melanoma 2; AIM2; Gm1313; Ifi210; ifi-210; Interferon-inducible protein 210; interferon-inducible protein AIM2; Interferon-inducible protein p210; PYHIN4; RP11-520H16.3
Gene Symbols AIM2
Host Species Mouse
Immunogen Recombinant human AIM2 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9447
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.