Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM2 Mouse anti-Human, Clone:7G12D1, Abnova™

Catalog No. 89333596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-596 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-596 Supplier Abnova Corporation Supplier No. MAB17173
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human AIM2.

AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2. [provided by RefSeq]

Specifications

Antigen AIM2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7G12D1
Conjugate Unconjugated
Description absent in melanoma 2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene AIM2
Gene Alias PYHIN4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-195 of human AIM2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 9447
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.