Learn More
Invitrogen™ AIM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA591996
Description
Positive Samples: HeLa, HepG2; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: HVKPEQKQMV AQQESIREGF QKRCLPVMVL KAKKPFTFET QEGKQEMFHA TVATEKEFFF VKVFNTLLKD KFIPKRIIII ARYYRHSGFL EVNSASRVLD AESDQKVNVP LNIIRKAGET PKINTLQTQP LGTIVNGLFV VQKVTEKKKN ILFDLSDNTG KMEVLGVRNE DTMKCKEGDK VRLTFFTLSK NGEKLQLTSG VHSTIKVI.
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.
Specifications
AIM2 | |
Polyclonal | |
Unconjugated | |
AIM2 | |
Absent in melanoma 2; AIM2; Gm1313; Ifi210; ifi-210; Interferon-inducible protein 210; interferon-inducible protein AIM2; Interferon-inducible protein p210; PYHIN4; RP11-520H16.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
304987, 383619, 9447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O14862, Q91VJ1 | |
AIM2 | |
A synthetic peptide corresponding to a sequence within amino acids 130-337 of human AIM2 (NP_0048241). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.