Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMP2 Antibody, Novus Biologicals™
SDP

Catalog No. p-200040011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433814 25 μL
NBP181574 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433814 Supplier Novus Biologicals Supplier No. NBP18157425UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AIMP2 Polyclonal specifically detects AIMP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AIMP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q13155
Gene Alias aminoacyl tRNA synthase complex-interacting multifunctional protein 2, aminoacyl tRNA synthetase complex-interacting multifunctional protein 2, ARS-interacting multi-functional protein 2, JTV1JTV-1, Multisynthase complex auxiliary component p38, multisynthetase complex auxiliary component p38, p38, PRO0992, Protein JTV-1
Gene Symbols AIMP2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:SVLGKDYGALKDIVINANPASPPLSLLVLHRLLCEHFRVLSTVHTHSSVKSVPENLLKCFGEQNKKQPRQDYQLGFTLIWKNVPKTQ
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7965
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.