Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984780
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4780 Supplier Bioss Supplier No. BS2509R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.

Specifications

Antigen AIMP2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AIMP2
Gene Accession No. Q13155
Gene Alias AA407136; AA407225; AIMP2; Aimp2(p38); Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; aminoacyl tRNA synthetase complex interacting multifunctional protein 2; aminoacyl tRNA synthetase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV1; JTV-1; Multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; P38; PRO0992; Protein JTV-1
Gene Symbols AIMP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AIMP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7965
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.