Learn More
AIMS™ ATS-3 General Rodent Tattoo System Needles
Click to view available options
Tip Style:
Heavy Duty Point
Standard Point
Ultra Fine Point
Description
Accessory for ATS-3 General Rodent Tattoo System
- Made of 401 stainless steel and silver solder
- Pre-sterilized and ready for tattooing
- Proper needles allows for humane and rapid animal tattooing
- Tails are not damaged
- 5 needles per pack
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
For Use With (Application) | Tattooing |
Type | Tattoo Needle |
For Use With (Equipment) | ATS-3 general rodent tattoo system |
Material | 401 Stainless Steel, Silver Solder |
Sterility | Sterile |
Tip Style | Heavy Duty Point |