Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIMS™ ATS-3 General Rodent Tattoo System Pigment
Click to view available options
Color:
Black
Green
Red
Capacity (English):
0.16 oz.
2 oz.
Capacity (Metric):
5 mL
59.14 mL
Description
- Safe
- Nontoxic
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
| Capacity (English) | 0.16 oz. |
| Capacity (Metric) | 5 mL |
| Color | Red |
| For Use With (Application) | In-vivo and in-vitro Toxicology Assay |
| Type | Tattoo Pigment |
| For Use With (Equipment) | ATS-3 general rodent tattoo system |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction