Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aiolos/IKZF3 Antibody (914821), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8625G
Description
Aiolos/IKZF3 Monoclonal antibody specifically detects Aiolos/IKZF3 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Aiolos/IKZF3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
914821 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
E. coli-derived recombinant human Aiolos/IKZF3, Met1-Gly54, Accession # Q9UKT9 | |
0.1 mL | |
Chromatin Research | |
22806 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction