Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aiolos/IKZF3 Antibody (OTI7E11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271698B
Description
Aiolos/IKZF3 Monoclonal specifically detects Aiolos/IKZF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Aiolos/IKZF3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
IKZF3 | |
Recombinant protein fragment corresponding to amino acids 1-301 of human Aiolos/IKZF3 (NP_036613) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI7E11 | |
Western Blot, Immunohistochemistry | |
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
Mouse | |
Affinity Purified | |
Chromatin Research | |
22806.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction