Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aiolos/IKZF3 Rabbit anti-Human, Mouse, Rat, Clone: 4V2Z2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
| Antigen | Aiolos/IKZF3 |
|---|---|
| Clone | 4V2Z2 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
Aiolos/IKZF3 Monoclonal antibody specifically detects Aiolos/IKZF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Aiolos/IKZF3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-509 of human Aiolos/IKZF3 (Q9UKT9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4V2Z2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 22806 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title