Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aiolos Monoclonal Antibody (8B2), eBioscience™, Invitrogen™

Catalog No. 14578937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-578-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-578-937 Supplier Invitrogen™ Supplier No. 14578937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This 8B2 monoclonal antibody reacts with mouse Aiolos, a 58-kDa member of the Ikaros family of zinc finger transcription factors. Expression of Aiolos is restricted to the lymphoid lineage, with B cells exhibiting the highest levels of expression, followed by NK and T cells. Studies demonstrate that Aiolos forms homodimers or heterodimerizes with Ikaros proteins. Moreover, Aiolos exhibits tyrosine phosphorylation and nuclear localization in response to IL-2 stimulation in T cells. Aiolos plays a role in IL-2-mediated anti-apoptosis by controlling Bcl-2 expression. Finally, Aiolos is involved in B cell development, with upregulation of this transcription factor observed at the pre-BI to pre-BII transition in response to pre-B cell receptor signaling. Applications Reported: This 8B2 antibody has been reported for use in immunoblotting/western blotting. Applications Tested: This 8B2 antibody has been tested by immunoblotting of reduced cell lysate prepared from mouse splenocytes. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aiolos
Applications Western Blot
Classification Monoclonal
Clone 8B2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ikzf3
Gene Accession No. O08900
Gene Alias 5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3
Gene Symbols Ikzf3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22780
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.