Learn More
Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$397.65
Specifications
Antigen | Aiolos |
---|---|
Clone | 8B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-578-982
![]() |
Invitrogen™
50578982 |
100 μg |
Each of 1
for $397.65
Save 25% Reg : |
![]() |
|||||
Description
This 8B2 MAb reacts with mouse Aiolos, a 58-kDa member of the Ikaros family of zinc finger transcription factors. Expression of Aiolos is restricted to the lymphoid lineage, with B cells exhibiting the highest levels of expression, followed by NK and T cells. Studies demonstrate that Aiolos forms homodimers or heterodimerizes with Ikaros proteins. Moreover, Aiolos exhibits tyrosine phosphorylation and nuclear localization in response to IL-2 stimulation in T cells. Aiolos plays a role in IL-2-mediated anti-apoptosis by controlling Bcl-2 expression. Finally, Aiolos is involved in B cell development, with upregulation of this transcription factor observed at the pre-BI to pre-BII transition in response to pre-B cell receptor signaling.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
Aiolos | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
O08900 | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
8B2 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Mouse | |
22780 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Ikzf3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.