Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIP/ARA9 Antibody (35-2), Novus Biologicals™
SDP

Catalog No. p-4978168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100127 0.1 mL
NB100127SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100127 Supplier Novus Biologicals Supplier No. NB100127
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIP/ARA9 Monoclonal specifically detects AIP/ARA9 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen AIP/ARA9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 35-2
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:200, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23702468), Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 21984905)
Formulation Tissue culture supernatant with 0.1% Sodium Azide
Gene Accession No. O00170
Gene Alias ARA9, FKBP16, FKBP37, SMTPHN, XAP2, XAP-2
Gene Symbols AIP
Host Species Mouse
Immunogen Bacterially expressed human AIP/ARA9 [UniProt# O00170]
Molecular Weight of Antigen 37 kDa
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Hypoxia, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 9049
Test Specificity Customers have reported success on rat lysate (see review).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.