Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AIP Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518715
Description
This antibody is predicted to react with canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
Specifications
AIP | |
Polyclonal | |
Unconjugated | |
Aip | |
AA408703; AH receptor-interacting protein; AIP; ARA9; aryl hydrocarbon receptor interacting protein; aryl-hydrocarbon receptor-interacting protein; AW476050; D19Bwg1412e; FKBP16; FKBP37; HBV X-associated protein 2; HBV-X associated protein; immunophilin homolog ARA9; Immunophilin XAP2; SMTPHN; XAP2; XAP-2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
11632, 9049 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O00170, O08915 | |
Aip | |
Synthetic peptide sequence (EARIRQKDEEDKAR) corresponding to the C-terminus amino acids of AIP (aa 310-323). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction