Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542294 Supplier Invitrogen™ Supplier No. PA542294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EMKYTSEGVR YFVDHNTRTT TFKDPRPGFE SGTKQGSPGA YDRSFRWKYH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%; Sheep: 100%; Zebrafish: 80%.

This gene encodes a member of the NEDD4-like protein family. The family of proteins is known to possess ubiquitin-protein ligase activity. The encoded protein contains 4 tandem WW domains. The WW domain is a protein motif consisting of 35 to 40 amino acids and is characterized by 4 conserved aromatic residues. The WW domain may mediate specific protein-protein interactions. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WWP2
Gene Accession No. O00308
Gene Alias 1300010O06Rik; AA690238; AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; AW554328; HECT-type E3 ubiquitin transferase WWP2; id:ibd1121; NEDD4-like E3 ubiquitin-protein ligase WWP2; wu:fo87e10; WW domain containing E3 ubiquitin protein ligase 2; WW domain-containing protein 2; WW domain-containing protein 4; WWP2; WWp2-like; zgc:154036
Gene Symbols WWP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human WWP2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11060
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.