Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100380 Supplier Thermo Scientific Supplier No. PA5100380
Add to Cart
Add to Cart

AIPL1 Polyclonal Antibody

AIPL1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen AIPL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AIPL1
Gene Accession No. Q924K1, Q9JLG9, Q9NZN9
Gene Alias A930007I01Rik; AIPL1; AIPL2; aryl hydrocarbon receptor interacting protein like 1; aryl hydrocarbon receptor interacting protein-like 1; aryl hydrocarbon receptor-interacting protein-like 1; aryl-hydrocarbon interacting protein-like 1; aryl-hydrocarbon-interacting protein-like 1; EGK_08083; LCA4
Gene Symbols AIPL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AIPL1(Accession Q9NZN9), corresponding to amino acid residues P339-H384.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114230, 23746, 59110
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.