Learn More
Air Systems™ Auto-Air™ Breather Box™ Air Filtration System Accessory, Particulate Filters
Shop All Air Systems International Products
Description
- For use with Auto-Air Breather Box air filtration system
- Particulate filter accessory
- Removes bulk water and particulates
- Use with Bb100-co series 100 cfm breather box
- Includes: A-style filter element and o-ring
Specifications
Specifications
| For Use With (Equipment) | BB100-CO Breather Box Series 100 cfm |
| Includes | A-style filter element and o-ring |
| Model | BB100-A |
| Product Type | Particulate Filter |