Learn More
AirClean™ Systems HEPA Filters
SDP
High efficiency particulate air (HEPA) filter, AirClean™ systems HEPA filters can be used with ductless fume hoods and drying cabinets.
$731.00 - $2184.00
Specifications
Filter Type HEPA
Product Type Filter
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
36-101-8805
SDP
View Documents
AirClean™ Systems
ACFHEPA18
Latent Print Dusting Station (DS200), Basic Forensic HEPA Workstations (AC648H), Advance Forensic Workstation (AC6400A)
Each for $731.00
Only null left
Add to Cart
36-101-8811
SDP
View Documents
AirClean™ Systems
ACFHEPA56
Horizontal Laminar Flow Workstation (AC5000HLF and UV)
Each for $2,184.00
Only null left
Add to Cart
Description
Description
  • High efficiency particulate air (HEPA) filters with 99.997% efficiency rating for collection at 0.3 microns
  • Used for containment of particles - toxic powders, asbestos, lead, fingerprint powders
Specifications
Specifications
HEPA
Filter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.