Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIRE Monoclonal Antibody (5H12), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Catalog No. 50246081
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-081 25 μg
50-246-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-246-081 Supplier Invitrogen™ Supplier No. 53593480
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This 5H12 monoclonal antibody reacts with mouse Autoimmune Regulator (AIRE). This 58-kDa transcription factor is highly expressed within the perinuclear region of mature medullary thymic epithelial cells (mTECs). However, expression of AIRE protein in the periphery remains unclear despite the detection of AIRE mRNA in the lymph node, lymphoid stromal cells, monocytes, and dendritic cells. Studies demonstrate that AIRE is involved in thymocyte development, in particular the regulation of tissue-specific antigen expression during negative selection. Other biological functions for AIRE include chemokine expression and antigen presentation. The 5H12 antibody recognizes mouse AIRE isoforms 1 and 2. Applications Reported: This 5H12 antibody has been reported for use in flow cytometric analysis and immunohistochemical staining of frozen tissue sections (IHC-F). Applications Tested: This 5H12 antibody has been tested by immunohistochemistry on fixed, frozen mouse thymus at less than or equal to 10 μg/mL. It is recommended that this antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

AIRE is a transcriptional regulator protein that binds to DNA as a dimer and tetramer, but not as a monomer, and may be involved in the regulation of gene expression. A nuclear protein, AIRE is a 545 amino acid protein with an HSR domain, two PHD type zinc fingers, and a SAND domain, and may be present in soluble high-molecular-weight complexes. It may form homodimer, homotetramer and also nuclear bodies and interacts with the transcriptional coactivator CBP and CREBBP. Three isoforms of AIRE are reported and one of them is predicted to be a candidate for nuclear-mediated decay (NMD). AIRE is involved in immune regulation and is known to mediate E3 ubiquitin ligase activity. Defects in AIRE are responsible for the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIRE
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 5H12
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Aire
Gene Accession No. Q9Z0E3
Gene Alias Aire; AIRE protein; AIRE1; APECED; APECED protein; APECED protein homolog; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune regulator; autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator AIRE1a; LOW QUALITY PROTEIN: autoimmune regulator; OTTHUMP00000109529; PGA1
Gene Symbols Aire
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11634
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2c κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.