Learn More
Invitrogen™ AIRE Polyclonal Antibody

Description
Positive Samples: MCF7, U-937, HT-29, OVCAR3, SW620, Mouse kidney, Mouse liver; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: HALADHDVVP EDKFQETLHL KEKEGCPQAF HALLSWLLTQ DSTAILDFWR VLFKDYNLER YGRLQPILDS FPKDVDLSQP RKGRKPPAVP KALVPPPRLP T.
Specifications
Specifications
| Antigen | AIRE |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Aire |
| Gene Accession No. | O43918, Q9Z0E3 |
| Gene Alias | Aire; AIRE protein; AIRE1; APECED; APECED protein; APECED protein homolog; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune regulator; autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator AIRE1a; LOW QUALITY PROTEIN: autoimmune regulator; OTTHUMP00000109529; PGA1 |
| Gene Symbols | Aire |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.