Learn More
MSA™ AirHawk™ II Self-Contained Breathing Apparatus: Advantage 4000 Model
Catalog No. 17200006 Shop All MSA Products
Change view
Click to view available options
Duration:
30 min.
45 min.
Harness Style:
Dupont™ Kevlar™ aramid fiber with chest strap, Speed-on™
Five strap, Rubber
Nylon with chest strap
Nylon with chest strap, polyester net
Nylon with chest strap, rubber
Speed-on™
Includes:
Advantage 4000 facepiece (medium Hycar rubber, polyester net head harness); Nylon harness and chest strap; Aluminum 30 min. cylinder; Hard case
Advantage 4000 facepiece (medium Hycar rubber, polyester net head harness); Nylon harness and chest strap; L30 stealth carbon; Hard case
Advantage 4000 facepiece, Polyester net head harness, Harness carrier: Nylon w/chest strap, Cylinder: carbon - 45 minutes
Advantage 4000 facepiece, Rubber net head harness, Harness carrier: Nylon w/chest strap; Cylinder: carbon - 30 minutes
Advantage 4000 facepiece; Polyester net head harness; Harness carrier: Nylon w/chest strap; Hard case
Facepiece: Ultra Elite; Rubber head harness
Facepiece: Ultra Elite; Rubber head harness; Hard case
Facepiece: Ultra Elite; Speed-On harness
Facepiece: Ultra Elite; Speed-On harness; Hard case
Facepiece: Ultra Elite; Speed-On harness; Harness carrier: Dupont™ Kevlar™ aramid fiber w/chest strap; Hard case
Harness carrier: Nylon w/chest strap
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration Harness Style Includes
17-200-006 45 min. Nylon with chest strap, polyester net Advantage 4000 facepiece, Polyester net head harness, Harness carrier: Nylon w/chest strap, Cylinder: carbon - 45 minutes
17-200-003 30 min. Speed-on™ Facepiece: Ultra Elite; Speed-On harness
17-200-001 30 min. Five strap, Rubber Facepiece: Ultra Elite; Rubber head harness
19-825-009 30 min. Nylon with chest strap, polyester net Advantage 4000 facepiece (medium Hycar rubber, polyester net head harness); Nylon harness and chest strap; Aluminum 30 min. cylinder; Hard case
17-200-002 30 min. Speed-on™ Facepiece: Ultra Elite; Speed-On harness; Hard case
19-825-011 30 min. Nylon with chest strap, polyester net Advantage 4000 facepiece (medium Hycar rubber, polyester net head harness); Nylon harness and chest strap; L30 stealth carbon; Hard case
17-200-005 45 min. Nylon with chest strap, polyester net Advantage 4000 facepiece; Polyester net head harness; Harness carrier: Nylon w/chest strap; Hard case
17-200-007 30 min. Nylon with chest strap, rubber Advantage 4000 facepiece, Rubber net head harness, Harness carrier: Nylon w/chest strap; Cylinder: carbon - 30 minutes
17-200-000 30 min. Five strap, Rubber Facepiece: Ultra Elite; Rubber head harness; Hard case
17-200-008 30 min. Nylon with chest strap Harness carrier: Nylon w/chest strap
17-200-004 45 min. Dupont™ Kevlar™ aramid fiber with chest strap, Speed-on™ Facepiece: Ultra Elite; Speed-On harness; Harness carrier: Dupont™ Kevlar™ aramid fiber w/chest strap; Hard case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Do not use for fire and potential firefighting-related applications. Unit is not NFPA approved for fire service.
Catalog No. 17-200-006 Supplier MSA™ Supplier No. 10116454
Do not use for fire and potential firefighting-related applications. Unit is not NFPA approved for fire service.
Lightweight, durable SCBA builds on the success of the established AirHawk air mask by providing options not typically offered in an industrial air mask
Available in both low- and high-pressure versions (2216 or 4500psig), it is suited for use in many industrial IDLH (immediately dangerous to life or health) applications.
Options tied to these basic components determine the features of the resulting SCBA configuration.
Facepiece
  • One-way check valve (to prevent exhaled air from entering the MMR) and a molded polycarbonate lens
  • Four-point silicone head harness for exceptional comfort and security; optional polyester net head harness also available
  • Adapters that allow it to convert to a twin-cartridge respirator (using Advantage cartridges) or an RD40 facepiece
First-Stage Regulator
  • PR-14 regulator is designed with the fewest parts in the industry (14) and can be serviced without special tools
  • PTFE fluoropolymer-coated anodized valve bodies minimize internal component friction and maximize service life
Mask-Mounted Regulator
  • Choose from push-to-connect or slide-to-connect versions of FireHawk™ MMR
  • Integrated shutoff prevents cylinder air loss when disconnecting regulator from facepiece
  • Has only 22 replaceable parts and can be serviced without special tools
Air Cylinder
  • Choose from three low-pressure (2216psig), 30-minute-rated cylinder options: carbon-wrapped, fiberglass hoop-wound, or all-aluminum construction
  • High-pressure (4500 psig) carbon-wrapped cylinders are available in 30-, 45-, or 60-minute durations
Airframe™ Carrier and Harness Assembly
  • Consists of an ergonomically designed composite backplate and choice of Nylon or Kevlar™ harness
  • Backplate is made from glass-fiber-reinforced thermoset plastic and features a spine strain relief; its key components are all easily detached without the use of special tools
  • Both harness styles feature a center-pull waist belt system as well as shoulder straps with easy-to-release friction buckles that positively retain the pull straps
  • Additional harness options include shoulder pads, lumbar pad and chest strap
End-of-Service-Time Indicator (EOSTI)
  • Audi-Larm™ air-actuated continuous warning bell automatically sounds when cylinder air supply reaches 25% of rated service life
  • Additional options include:
    • Air-line port—Air mask can be outfitted with a second port designed to accept connection to an external air source such as a compressor or cascade system
    • Heads-up display—Wireless display is available on Ultra Elite facepiece and consists of integrated transmitter and receiver; display lights show remaining volume of cylinder air
    • Rapid Intervention Crew (RIC) system—Enables quick refilling from compressor or cascade system as well as transfill between two SCBAs
    • Emergency egress rescue belt—NIOSH-certified emergency egress system is integral to SCBA waist belt and comes complete with Kevlar line and rappelling hardware
    • Shoulder-mounted Quick Fill System EBS—Offers SCBA cylinder refill from a mobile compressor or cascade system as well as transfill between two SCBAs
    • ICM™ Tx Integrated PASS/HUD transmitter—Combines Personal Alert Safety System (PASS) and HUD transmitter within a single housing; easily replaced wireless system eliminates snag hazards
    Order Info
    All AirHawk II SCBAs are assembled-to-order utilizing the variety of options detailed in the ordering matrix below. This allows each customer to custom-configure an air mask with only the options they desire. For assistance in using the matrix or to place an order from it, call your Fisher Safety Customer Service Representative at 1-800-772-6733.
    Certifications
    NIOSH approved.
    Compliance
    Non-NFPA compliant.
    Specifications
    Certifications/Compliance NIOSH, Non-NFPA
    Product Type SCBA Assembly
    Duration 45 min.
    Facepiece Type Advantage 4000
    For Use With (Application) Suited for use in many industrial IDLH (immediately dangerous to life or health) applications
    Harness Style Nylon with chest strap, polyester net
    Includes Advantage 4000 facepiece, Polyester net head harness, Harness carrier: Nylon w/chest strap, Cylinder: carbon - 45 minutes
    Material Medium Hycar™