Learn More
AirClean™ Systems AirMax™ Polypropylene Total Exhaust Fume Hoods
Standard bypass design of the AirClean Systems AirMax™ Polypropylene Total Exhaust Fume Hood exhausts constant volume of air, regardless of sash position.
$12229.00 - $19994.00
| Catalog Number | Mfr. No. | Dimensions (D x W x H) Interior | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (D x W x H) Interior | Price | Quantity & Availability | |||||
36-100-0046
![]() |
AirClean™ Systems
AC5030TE |
21 x 49 x 32 in. (53.3 x 124.4 x 81.2 cm) |
Each for $13,224.00
|
|
|||||
36-100-0044
![]() |
AirClean™ Systems
AC4030TE |
21 x 37 x 32 in. (53.3 x 94 x 81.2 cm) |
Each for $12,229.00
|
|
|||||
36-100-0048
![]() |
AirClean™ Systems
AC8030TE |
21 x 85 x 32 in. (53.3 x 216 x 81.2 cm) |
Each for $19,994.00
|
|
|||||
36-100-0047
![]() |
AirClean™ Systems
AC6030TE |
21 x 183 x 32 in. (53.3 x 155 x 81.2 cm) |
Each for $14,398.00
|
|
|||||
36-100-0045
![]() |
AirClean™ Systems
AC3030TE |
21 x 25 x 32 in. (53.3 x 63.5 x 81.2 cm) | N/A | N/A | N/A | ||||
Description
- Manufactured from all-white, flame-retardant polypropylene
- NO RUST- all contact surfaces are constructed of polypropylene
- Microprocessor controller monitors face velocity and displays the real-time airflow on the LCD screen in linear feet per minute
- Vertical sliding safety glass sash
- Built-in vapor-proof fluorescent light
- Easy-access utilities panel for operation and maintenance
- Non-hydroscopic, will not absorb moisture
- Large volume spill basin with solid work surface
Safety Features:
- Advanced airfoil design that is important to the overall safety and performance of the fume hood
- Rolled entry and advanced air foil design prevents reverse flow and eddy currents from escaping the fume hood
- Double sidewall construction allows mounting of services and outlets on the posts
- Effective spill containment- Removable work surface exposes spill basin that may be fitted with drain for disposal of any spills
- Removable work surface exposes spill basin that may be fitted with drain for disposal of any spills
- Rear baffle extends below work surface to effectively remove any heavy gases or evaporation from a spill
- Can be fitted with wet scrubber system to remove high volumes of water soluble acids and solvents
- Auto dosing system for neutralization of acids
- Polycarbonate sash for HF applications
- Polypropylene blowers
- Variable speed driver to make a true VAV system
- Polypropylene base cabinets
Specifications
| Total Exhaust Fume Hood | |
| Polypropylene |