Learn More
Air-Tite™ All-Plastic Henke-Ject™ Syringes
Catalog No. 1481741 Shop All Air-Tite Products
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
50 mL
Graduations:
0.05 cc
0.1 cc
0.2 cc
0.5 cc
1 cc
2 cc
Connection Type:
Catheter
Centric Luer
Centric Luer Slip
Eccentric Catheter
Eccentric Luer Tip
Leur Slip
Luer Lock
Luer Tip
This item is not returnable. View return policy
This item is not returnable. View return policy
Requires
Purchase needles separately.
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Henke-Ject™ syringes are available compatible with a variety of needle styles. Needles are sold separately.
These Air-Tite, all plastic syringes are available compatible with a variety of needle styles. (Needles are sold separately.) Ideal for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures.
  • Needle tips not included and must be purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
  • Luer Tip, Luer-Slip, or catheter tip compatibility
  • Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
  • Positive safety stop prevents accidental spills
  • FDA registered
  1. Dose saver design with low dead space plug on the piston minimizes waste.
  2. Catheter syringe has a large opening similar to a nozzle and is usable with catheters.
Specifications
Certifications/Compliance FDA, CE, ISO 9001, DEHP Free
Material Plastic
For Use With (Application) For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures
Latex Free Yes
Product Type Henke-Ject™ Syringe
Graduations 1 cc
Volume (Metric) 30 mL
Needle Point Style Cone
Connection Type Luer Tip
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.