Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTRAX™ Airug™ Mat

Click to view available options
Color:
Black
Black/Yellow
Gray
Description
- PVC closed cell foam construction relieves leg and back discomfort while providing a barrier to cold floors
- Basic anti-fatigue protection provided
- Beveled edges on all four sides
Specifications
Specifications
Width (English) | 60 ft. |
Thickness (English) | 3/8 in. |
Color | Black |
Length (English) | 3 ft. |
Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction