Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AJAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578684 Supplier Invitrogen™ Supplier No. PA578684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: rat liver Predicted Reactivity: Human (100%), Mouse (91%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AJAP1, Adherens Junction Associated Protein 1, plays a key role in cell adhesion and cell migration. It is a transmembrane protein found on the basolateral surface of epithelium and expressed by multiple types of epithelium. AJAP1 interacts with E-Cadherin and appears to promote the internalization of E-Cadherin upon growth factor stimulation. This facilitates the dissolution of adherens junctions with an increase in cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AJAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene AJAP1
Gene Accession No. Q4W8E7, Q9UKB5
Gene Alias adherens junction associated protein 1; adherens junction-associated protein 1; adherens junctions associated protein 1; AJAP1; Gm573; likely ortholog of H. sapiens transmembrane protein SHREW1; membrane protein shrew-1; MOT8; Neumo48; RP3426F10\.1; SHREW1; SHREW-1; transmembrane protein SHREW1
Gene Symbols AJAP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human AJAP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55966, 687031
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.