Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AJAP1 Polyclonal Antibody, Bioss
SDP

Catalog No. 502280620
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0620 Supplier Bioss Supplier No. BS22384R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AJAP1 Polyclonal antibody specifically detects AJAP1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen AJAP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene AJAP1
Gene Accession No. A2ALI5, Q4W8E7, Q9UKB5
Gene Alias adherens junction associated protein 1; adherens junction-associated protein 1; adherens junctions associated protein 1; AJAP1; Gm573; likely ortholog of H. sapiens transmembrane protein SHREW1; membrane protein shrew-1; MOT8; Neumo48; RP3426F10\.1; SHREW1; SHREW-1; transmembrane protein SHREW1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AJAP1
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230959, 55966, 687031
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.