Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AJAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA538044
Description
A recommended positive control is rat liver tissue lysate.
AJAP1, Adherens Junction Associated Protein 1, plays a key role in cell adhesion and cell migration. It is a transmembrane protein found on the basolateral surface of epithelium and expressed by multiple types of epithelium. AJAP1 interacts with E-Cadherin and appears to promote the internalization of E-Cadherin upon growth factor stimulation. This facilitates the dissolution of adherens junctions with an increase in cell motility.
Specifications
AJAP1 | |
Polyclonal | |
Unconjugated | |
AJAP1 | |
adherens junction associated protein 1; adherens junction-associated protein 1; adherens junctions associated protein 1; AJAP1; Gm573; likely ortholog of H. sapiens transmembrane protein SHREW1; membrane protein shrew-1; MOT8; Neumo48; RP3426F10\.1; SHREW1; SHREW-1; transmembrane protein SHREW1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230959, 55966, 687031 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A2ALI5, Q4W8E7, Q9UKB5 | |
AJAP1 | |
18 amino acid peptide near the carboxy terminus of human AJAP1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction