Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AJUBA Monoclonal Antibody (21G4)
GREENER_CHOICE

Catalog No. PIMA518087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518087 Supplier Invitrogen™ Supplier No. MA518087
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This target has a predicted molecular weight of 57 kDa.

AJUBA (Ajuba LIM Protein) is a Protein Coding gene. Diseases associated with AJUBA include Wilson-Turner X-Linked Mental Retardation Syndrome and Syndromic X-Linked Intellectual Disability. Among its related pathways are Hippo signaling pathway and Adhesion. Gene Ontology (GO) annotations related to this gene include chromatin binding and actin filament binding. An important paralog of this gene is LIMD1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AJUBA
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 21G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene AJUBA
Gene Accession No. Q96IF1
Gene Alias AJUBA; ajuba homolog; ajuba LIM protein; Ajuba protein; JUB; jub, ajuba homolog; LIM domain-containing protein ajuba; protein ajuba
Gene Symbols AJUBA
Host Species Mouse
Immunogen Full length Human Ajuba fused to maltose binding protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.