Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AJUBA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109373 Supplier Invitrogen™ Supplier No. PA5109373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERLGEKASR LLEKFGRRKG ESSRSGSDGT PGPGKGRLSG LGGPRKSGPR GATGGPGDEP LEPAREQGSL DAERNQRGSF EAPRYEGSFP AGPPPTRALP.

AJUBA (Ajuba LIM Protein) is a Protein Coding gene. Diseases associated with AJUBA include Wilson-Turner X-Linked Mental Retardation Syndrome and Syndromic X-Linked Intellectual Disability. Among its related pathways are Hippo signaling pathway and Adhesion. Gene Ontology (GO) annotations related to this gene include chromatin binding and actin filament binding. An important paralog of this gene is LIMD1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AJUBA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AJUBA
Gene Accession No. Q5U2Z2, Q91XC0, Q96IF1
Gene Alias AJUBA; ajuba homolog; ajuba LIM protein; Ajuba protein; JUB; jub, ajuba homolog; LIM domain-containing protein ajuba; protein ajuba
Gene Symbols AJUBA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Ajuba (NP_1162651).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16475, 84962, 85265
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.