Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AK 7
GREENER_CHOICE

Catalog No. 475410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-541-0 10 mg
47-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-541-0 Supplier Tocris Bioscience™ Supplier No. 4754/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective SIRT2 inhibitor; brain penetrant

Selective SIRT2 inhibitor (IC50 = 15.5 μM); displays no effect on SIRT1 or SIRT3. Decreases neuronal cholesterol levels; improves motor function and ameliorates brain atrophy in a mouse model of Huntington's disease. Brain penetrant.

Chemical Identifiers

CAS 420831-40-9
Molecular Formula C19H21BrN2O3S

Specifications

Quantity 10 mg
Formula Weight 437.35
Purity >98%
Molecular Weight (g/mol) 437.35
Chemical Name or Material AK 7
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.