Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AK Protective Benchtop Shields

Catalog No. 17987140C
Encompass
Click to view available options
Width (Metric):
30.4 cm
40.6 cm
Thickness (Metric):
0.63 cm
0.95 cm
Width (English):
12 in.
16 in.

These durable acrylic shields are ideal for use with blood, liquids, culture specimens, or other hazardous materials. The angled top protects the face against splash and flying particles, while the patented curved-side design allows more natural arm movement, so workers can more easily reach the work surface.Shields are available in two thicknesses: 3/8 and 1/4 in. (0.95 and 0.6cm). The 3/8 in. thick (0.95cm) shield offers protection against radiation exposure when used with low-level beta-emitting isotopes such as 32P. The 1/4 in. thick (0.6cm) shield should be used for splash protection only. Both shields offer a choice of two work surface sizes: 12L × 12 in.W (31 × 31cm) or 12L × 16 in.W (31 × 41cm). All shields are 20 in. high (51cm).

Specifications

Height (Metric) 50.8 cm
Width (English) 16 in.
Width (Metric) 40.6 cm
Height (English) 20 in.
Thickness (Metric) 0.63 cm
For Use With (Equipment) For use with blood, liquids, culture specimens, or other hazardous materials
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.