Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AK2 Antibody (AT7E7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250575H
Description
AK2 Monoclonal specifically detects AK2 in Human samples. It is validated for Western Blot, ELISA.Specifications
AK2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AK2 | |
Recombinant human AK2 (1-239aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
AT7E7 | |
Western Blot, ELISA | |
adenylate kinase 2, adenylate kinase 2, mitochondrial, adenylate kinase isoenzyme 2, mitochondrial, ADK2, AK 2, ATP-AMP transphosphorylase 2, EC 2.7.4, EC 2.7.4.3 | |
Mouse | |
Protein A purified | |
RUO | |
204 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction