Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AK3 Antibody (OTI6G4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271874F
Description
AK3 Monoclonal specifically detects AK3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
AK3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
AK3 | |
Full length recombinant protein of human AK3 (NP_057336) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI6G4 | |
Flow Cytometry, Immunohistochemistry | |
Adenylate kinase 3 alpha-like 1, adenylate kinase 3 like 1, adenylate kinase 3adenylate kinase 6, AK 3, AK6, AKL3L, AKL3L1, EC 2.7.4, EC 2.7.4.10, FIX, GTP:AMP phosphotransferase, mitochondrial | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
50808.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction