Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118932
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118932 200 μL
PIPA5122269 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118932 Supplier Invitrogen™ Supplier No. PA5118932
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AK3 Polyclonal Antibody for Western Blot

This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AK3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AK3
Gene Accession No. Q9UIJ7, Q9WTP7
Gene Alias AA407498; adenylate kinase 3; adenylate kinase 3 {ECO:0000255; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255; adenylate kinase 6, adenylate kinase 3 like 1; AI506714; AK 3; AK 3 {ECO:0000255; AK3; AK-3; Ak3l; AK3L1; AK4; AK6; AKL3L; AKL3L1; FIX; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial; HAMAP-Rule:MF_03169}
Gene Symbols AK3
Host Species Rabbit
Immunogen Recombinant protein Adenylate Kinase 3. The antigen corresponds to amino acid range 8-227 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100155691, 50808, 56248
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.