Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AK3L1 Antibody (OTI3B1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270129MFV500
Description
AK3L1 Monoclonal antibody specifically detects AK3L1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
AK3L1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI3B1 | |
Flow Cytometry | |
adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase | |
Full length human recombinant protein of human AK4(NP_037542) produced in HEK293 cell. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
205 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction