Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104541 Supplier Invitrogen™ Supplier No. PA5104541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KAD6.

Broad-specificity nucleoside monophosphate (NMP) kinase that catalyzes the reversible transfer of the terminal phosphate group between nucleoside triphosphates and monophosphates. AMP and dAMP are the preferred substrates, but CMP and dCMP are also good substrates. IMP is phosphorylated to a much lesser extent. All nucleoside triphosphates ATP, GTP, UTP, CTP, dATP, dCTP, dGTP, and TTP are accepted as phosphate donors. CTP is the best phosphate donor, followed by UTP, ATP, GTP and dCTP. May have a role in nuclear energy homeostasis. Has also ATPase activity. May be involved in regulation of Cajal body (CB) formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AK6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AK6
Gene Accession No. Q5EB68, Q8VCP8, Q9Y3D8
Gene Alias 2810046E22Rik; 4921516M08Rik; AD-004; adenylate kinase 6; adenylate kinase isoenzyme 6; adenylate kinase isoenzyme 6 {ECO:0000255; Adrenal gland protein AD-004; AK/ATPase; AK/ATPase {ECO:0000255; AK6; AK6 {ECO:0000255; ATP-AMP transphosphorylase 6; CGI-137; CINAP; CIP; coilin interacting nuclear ATPase protein; coilin interacting protein; coilin-interacting nuclear ATPase protein; coilin-interacting nuclear ATPase protein {ECO:0000255; Dual activity adenylate kinase/ATPase; dual activity adenylate kinase/ATPase {ECO:0000255; HAMAP-Rule:MF_03173}; hCINAP
Gene Symbols AK6
Host Species Rabbit
Immunogen A synthesized peptide derived from human AK6(Accession Q9Y3D8), corresponding to amino acid residues G15-L65.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102157402, 102216272, 102238592
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.