Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAIN1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281698AF750
This item is not returnable.
View return policy
Description
AKAIN1 Polyclonal antibody specifically detects AKAIN1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
AKAIN1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
C18orf42, C18orf42 chromosome 18 open reading frame 42 | |
AKAIN1 antibody was raised against a 16 amino acid peptide from near the center of human AKAIN1. The immunogen is located within the last 50 amino acids of AKAIN1. Amino Acid Squence: VSQESQRREERISDNR | |
0.1 mL | |
Neuroscience | |
642597 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction