Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $666.47
Specifications
| Antigen | AKAP1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
AKAP1 Polyclonal specifically detects AKAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| AKAP1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 8165 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EHVLELENSKGPSLASLEGEEDKGKSSSSQVVGPVQEEEYVAEKLPSRFIESAHTELAKDDAAPAPPVADAKAQDRGVEGELGNEESLDRNEEGLDRNEEGLDRNEESLDRNEEGLDRNEEIKRAAFQIISQVISEATEQV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| A kinase (PRKA) anchor protein 1, a kinase anchor protein 1, mitochondrial, AKAP121, AKAP149AKAP 149, AKAP84, A-kinase anchor protein 1, mitochondrial, A-kinase anchor protein 149 kDa, A-kinase anchor protein, 149kD, D-AKAP1, D-AKAP-1, Dual specificity A-kinase-anchoring protein 1, dual-specificity A-kinase anchoring protein 1, MGC1807, PRKA1, protein kinase A anchoring protein 1, protein kinase A1, Protein kinase A-anchoring protein 1, protein kinase anchoring protein 1, S-AKAP84, SAKAP84AKAP, Spermatid A-kinase anchor protein 84 | |
| AKAP1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title