Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571818
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1818 10 μL
50-157-1817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1818 Supplier Bethyl Laboratories, Inc Supplier No. IHC00281T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Northern white-cheeked gibbon

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein binds to type I and type II regulatory subunits of PKA and anchors them to the mitochondrion. This protein is speculated to be involved in the cAMP-dependent signal transduction pathway and in directing RNA to a specific cellular compartment.

Specifications

Antigen AKAP1
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene AKAP1
Gene Accession No. Q92667
Gene Alias AKAP, AKAP121, AKAP149, AKAP84, D-AKAP1, PPP1R43, PRKA1, SAKAP84, A-kinase anchor protein 1, mitochondrial, A-kinase anchor protein 149 kDa, AKAP 149, D-AKAP-1, dual-specificity A-kinase anchoring protein 1, protein kinase A anchoring protein 1, protein kinase A1, protein phosphatase 1, regulatory subunit 43, spermatid A-kinase
Gene Symbols AKAP1
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8165
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.