Learn More
AKAP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301379A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein binds to type I and type II regulatory subunits of PKA and anchors them to the mitochondrion. This protein is speculated to be involved in the cAMP-dependent signal transduction pathway and in directing RNA to a specific cellular compartment.Specifications
| AKAP1 | |
| Polyclonal | |
| Unconjugated | |
| AKAP1 | |
| AKAP, AKAP121, AKAP149, AKAP84, D-AKAP1, PPP1R43, PRKA1, SAKAP84, A-kinase anchor protein 1, mitochondrial, A-kinase anchor protein 149 kDa, AKAP 149, D-AKAP-1, dual-specificity A-kinase anchoring protein 1, protein kinase A anchoring protein 1, protein kinase A1, protein phosphatase 1, regulatory subunit 43, spermatid A-kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8165 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q92667 | |
| AKAP1 | |
| Between 375 and 425 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.